missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-3,3-dimethylisoquinoline, TRC

Product Code. 30135538
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135538 100 mg 100mg
30008089 10 mg 10mg
30005400 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135538 Supplier Toronto Research Chemicals Supplier No. TRCD450720100MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28460-55-1
Molecular Formula C11H13N
Molecular Weight (g/mol) 159.23
Synonym 3,4-Dihydro-3,3-dimethyl-isoquinoline
IUPAC Name 3,3-dimethyl-4H-isoquinoline
SMILES CC1(C)Cc2ccccc2C=N1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C11H13N/c1-11(2)7-9-5-3-4-6-10(9)8-12-11/h3-6,8H,7H2,1-2H3
Formula Weight 159.1048
Chemical Name or Material 3,4-Dihydro-3,3-dimethylisoquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.