missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxy-3-cyclobutene-1,2-dione, TRC

Product Code. 30376454
Change view
Click to view available options
Quantity:
5 g
Product Code. Quantity
30376454 5 g
1 options
This item is not returnable. View return policy

Product Code. 30376454

Brand: Toronto Research Chemicals TRCD4574555G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 5222-73-1
Molecular Formula C6H6O4
Quantity 5 g
Synonym 1,2-Dimethoxy-1-cyclobutene-3,4-dione, 1,2-Dimethoxy-3,4-cyclobutenedione, 1,2-Dimethoxy-3,4-dioxocyclobutene, 1,2-Dimethoxycyclobutenedione, 3,4-Dimethoxy-1,2-cyclobutenedione, Dimethyl quadratate, Dimethyl squarate
InChI Formula InChI=1S/C6H6O4/c1-9-5-3(7)4(8)6(5)10-2/h1-2H3
SMILES COC1=C(OC)C(=O)C1=O
Molecular Weight (g/mol) 142.11
Formula Weight 142.0266
Chemical Name or Material 3,4-Dimethoxy-3-cyclobutene-1,2-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.