missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxy-L-phenylalanine, 97%

Product Code. 15470108
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15470108 1 g 1g
15480108 5 g 5g
15490108 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15470108 Supplier Thermo Scientific Alfa Aesar Supplier No. H53368.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 32161-30-1 | C11H15NO4 | 225.244 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32161-30-1
Molecular Formula C11H15NO4
Molecular Weight (g/mol) 225.244
MDL Number MFCD01321274
InChI Key VWTFNYVAFGYEKI-QMMMGPOBSA-N
Synonym 3,4-dimethoxy-l-phenylalanine, s-2-amino-3-3,4-dimethoxyphenyl propionic acid, 3-3,4-dimethoxyphenyl-l-alanine, l-3,4-dimethoxyphenylalanine, l-3,4-dimethoxyphenyl alanine, tyrosine, 3-methoxy-o-methyl, 3,4-dimethoxyphenylalanine, l, unii-gc6uei64ec, s-2-amino-3-3,4-dimethoxy-phenyl-propionic acid, l-tyrosine, 3-methoxy-o-methyl
PubChem CID 10632946
IUPAC Name (2S)-2-amino-3-(3,4-dimethoxyphenyl)propanoic acid
SMILES COC1=C(C=C(C=C1)CC(C(=O)O)N)OC

Specifications

Melting Point 254°C to 257°C
Quantity 1 g
Formula Weight 225.24
Optical Rotation −5° (c=4 in 1M Hydrochloric Acid)
Sensitivity Air Sensitive
Percent Purity 97%
Chemical Name or Material 3,4-Dimethoxy-L-phenylalanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.