missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxy-N-methylbenzylamine, 97%, ACROS Organics™

Brand:  Acros Organics 439680050

Code : 29

Additional Details : CAS Number : 63-64-9 Weight : 0.05500kg Transport : UN number : 2735 Chem class : 8 Pack group : III


Quantity 5g

 View more versions of this product

Product Code. 11955311

  • 119.00 € / 5g

Estimated Shipment: 05-07-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

63-64-9
181.24
3,4-dimethoxybenzyl methylamine, n-methyl-3,4-dimethoxybenzylamine, 1-3,4-dimethoxyphenyl-n-methylmethanamine, 3,4-dimethoxyphenyl methyl methyl amine, 3,4-dimethoxy-n-methylbenzylamine, 3,4-dimethoxy-benzyl-methyl-amine, benzenemethanamine, 3,4-dimethoxy-n-methyl, 1-3,4-dimethoxyphenyl-n-methyl-methanamine, 3,4-dimethoxyphenyl methyl methylamine, 3,4-dimethoxybenzyl-n-methylamine
1-(3,4-dimethoxyphenyl)-N-methylmethanamine
C10H15NO2
XQODFBIAQVJQHF-UHFFFAOYSA-N
592161
CNCC1=CC(=C(C=C1)OC)OC

Specifications

3, 4-Dimethoxy-N-methylbenzylamine
Authentic
5g
96.0
96% min. (GC)
3,4-dimethoxybenzyl methylamine, n-methyl-3,4-dimethoxybenzylamine, 1-3,4-dimethoxyphenyl-n-methylmethanamine, 3,4-dimethoxyphenyl methyl methyl amine, 3,4-dimethoxy-n-methylbenzylamine, 3,4-dimethoxy-benzyl-methyl-amine, benzenemethanamine, 3,4-dimethoxy-n-methyl, 1-3,4-dimethoxyphenyl-n-methyl-methanamine, 3,4-dimethoxyphenyl methyl methylamine, 3,4-dimethoxybenzyl-n-methylamine
CNCC1=CC(=C(C=C1)OC)OC
181.24
181.24
97%
110°C
Brown to Colorless
63-64-9
100.0
C10H15NO2
XQODFBIAQVJQHF-UHFFFAOYSA-N
1-(3,4-dimethoxyphenyl)-N-methylmethanamine
592161
Liquid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3,4-Dimethoxy-N-methylbenzylamine

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS Documents
Certificates