missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxyphenylmagnesium bromide, 0.5M in THF

Product Code. 15451446
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15451446 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15451446 Supplier Thermo Scientific Alfa Aesar Supplier No. H54654.AE
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 89980-69-8 | C8H9BrMgO2 | 241.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a pharmaceutical intermediate.

Solubility
Reacts with water.

Notes
Air and moisture sensitive. Air, oxidizing agents, heat are incompatible. Store under cool place.
TRUSTED_SUSTAINABILITY

Specifications

CAS 89980-69-8
Molecular Formula C8H9BrMgO2
MDL Number MFCD01311476
Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive; May form a precipitate
Synonym 3,4-dimethoxyphenylmagnesium bromide, 3,4-dimethoxyphenyl magnesium bromide, 3,4-dimethoxyphenylmagnesium bromide solution, bhtpzwcaihfrsy-uhfffaoysa-m, 3,4-dimethoxyphenylmagnesium bromide, 0.5m in thf, 3,4-dimethoxyphenylmagnesium bromide 0.5 m in tetrahydrofuran, 3,4-dimethoxyphenylmagnesium bromide solution, 0.5 m in thf, grignard reagent
Solubility Information Reacts with water.
InChI Key BHTPZWCAIHFRSY-UHFFFAOYSA-M
SMILES COC1=CC=C([Mg]Br)C=C1OC
IUPAC Name magnesium;1,2-dimethoxybenzene-5-ide;bromide
Molecular Weight (g/mol) 241.37
PubChem CID 11010104
Formula Weight 241.37
Concentration or Composition (by Analyte or Components) 0.5M in THF
Chemical Name or Material 3,4-Dimethoxyphenylmagnesium bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.