missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethyl-1-pentyn-3-ol, 94%

Product Code. 11478737
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11478737 25 g 25g
11468737 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11478737 Supplier Thermo Scientific Alfa Aesar Supplier No. A19906.14
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1482-15-1 | C7H12O | 112.17 g/mol

3,4-Dimethyl-1-pentyn-3-ol is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethyl-1-pentyn-3-ol is used as pharmaceutical intermediates.

Solubility
Fully miscible in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1482-15-1
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.17
MDL Number MFCD00039845
InChI Key DZNLEQBXXLGELU-UHFFFAOYNA-N
PubChem CID 95483
IUPAC Name 3,4-dimethylpent-1-yn-3-ol
SMILES CC(C)C(C)(O)C#C

Specifications

Density 0.87
Boiling Point 133°C
Flash Point 40°C (104°F)
Quantity 25 g
UN Number UN1987
Synonym 3,4-dimethyl-1-pentyn-3-ol, 1-pentyn-3-ol, 3,4-dimethyl, acmc-1bzc3, 2-isopropyl-3-butyne-2-ol, 1-pentyn-3-ol,4-dimethyl, 3,4-dimethyl-pent-1-yn-3-ol, dznleqbxxlgelu-uhfffaoysa, 1-pentyn-3-ol,3,4-dimethyl, 1,2-dimethyl-1-hydroxypropyl acetylene
Solubility Information Fully miscible in water.
Formula Weight 112.17
Percent Purity 94%
Chemical Name or Material 3,4-Dimethyl-1-pentyn-3-ol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.