missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylanisole, 99%

Product Code. 11479747
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479747 50 g 50g
11489747 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11479747 Supplier Thermo Scientific Alfa Aesar Supplier No. B22735.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4685-47-6 | C9H12O | 136.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4685-47-6
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00008396
InChI Key LVUBSVWMOWKPDJ-UHFFFAOYSA-N
Synonym 3,4-dimethylanisole, 4-methoxy-1,2-dimethyl-benzene, 4-methoxy-o-xylene, benzene, 4-methoxy-1,2-dimethyl, 3,4-dimethyl anisole, 1,2-dimethyl-4-methoxybenzene, 3,4-dimethylphenyl methyl ether, acmc-20anel, pubchem5361
PubChem CID 78411
IUPAC Name 4-methoxy-1,2-dimethylbenzene
SMILES CC1=C(C=C(C=C1)OC)C

Specifications

Density 0.975
Boiling Point 200°C to 201°C
Quantity 50 g
Refractive Index 1.5175
Beilstein 2040907
Formula Weight 136.19
Percent Purity 99%
Chemical Name or Material 3,4-Dimethylanisole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.