missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dimethylanisole, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | 3,4-Dimethylanisole |
| CAS | 4685-47-6 |
| Molecular Formula | C9H12O |
| Quantity | 25 g |
| Synonym | 4-Methoxy-1,2-dimethylbenzene, 1,2-Dimethyl-4-methoxybenzene1-Methoxy-3,4-dimethylbenzene, 2-Methyl-4-methoxytoluene |
| InChI Formula | InChI=1S/C9H12O/c1-7-4-5-9(10-3)6-8(7)2/h4-6H,1-3H3 |
| SMILES | COc1ccc(C)c(C)c1 |
| IUPAC Name | 4-methoxy-1,2-dimethylbenzene |
| Molecular Weight (g/mol) | 136.19 |
| Formula Weight | 136.0888 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction