missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylhexane, 99%, Thermo Scientific™

Product Code. 13427756 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 mL
Packaging:
Glass bottle
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13427756 1 mL Glass bottle 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 13427756 Supplier Thermo Scientific Acros Supplier No. 443920010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Specifications

CAS 583-48-2
Density 0.7200g/mL
Boiling Point 119.0°C
Flash Point 6°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C8H18
Linear Formula CH3CH2CH(CH3)CH(CH3)CH2CH3
Quantity 1 mL
Specific Gravity 0.72
Synonym hexane, 3,4-dimethyl, 3,4-dimethyl hexane, acmc-1aquh, c2h5ch ch3 ch ch3 c2h5, 3,4-dimethylhexane, analytical standard, 3,4-dimethylhexane, mixture of +/-and meso
InChI Key RNTWWGNZUXGTAX-UHFFFAOYSA-N
SMILES CCC(C)C(C)CC
IUPAC Name 3,4-dimethylhexane
Molecular Weight (g/mol) 114.23
PubChem CID 11412
Formula Weight 114.23
Percent Purity 99%
Chemical Name or Material 3, 4-Dimethylhexane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3,4-Dimethylhexane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.