missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylphenylacetic acid, 98%

Product Code. 15469728
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15469728 5 g 5g
15459728 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15469728 Supplier Thermo Scientific Alfa Aesar Supplier No. H63178.06
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 17283-16-8 | C10H11O2 | 163.20 g/mol

3,4-Dimethylphenylacetic acid, is is used as an intermediate in organic synthesis. It is also used as a fine chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethylphenylacetic acid, is is used as an intermediate in organic synthesis. It is also used as a fine chemical intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17283-16-8
Molecular Formula C10H11O2
Molecular Weight (g/mol) 163.20
MDL Number MFCD02664684
InChI Key OTTPBKINJOYJGW-UHFFFAOYSA-M
PubChem CID 296013
IUPAC Name 2-(3,4-dimethylphenyl)acetic acid
SMILES CC1=CC=C(CC([O-])=O)C=C1C

Specifications

Quantity 5 g
Physical Form Solid
Synonym 2-3,4-dimethylphenyl acetic acid, 3,4-dimethylphenylacetic acid, 3,4-dimethyl-phenyl-acetic acid, 3,4-dimethylphenylaceticacid, 3,4-dimethyphenylacetic acid, 3,4-dimethylphenyl acetic acid, benzeneacetic acid, 3,4-dimethyl, 3,4-dimethylbenzenacetic acid, pubchem10440, benzeneacetic acid,3,4-dimethyl
Solubility Information Slightly soluble in water.
Formula Weight 164.2
Percent Purity 98%
Chemical Name or Material 3,4-Dimethylphenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.