missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylphenylzinc iodide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Product Code. 15485325
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
This item is not returnable. View return policy

Product Code. 15485325

Brand: Thermo Scientific Alfa Aesar H58165.AD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3,4-Dimethylphenylzinc iodide is used as pharmaceutical intermediates.

Solubility
Reacts with water.

Notes
Store at 4°C. Incompatible with oxidizing agents, acid chlorides, acids. Store under dr
TRUSTED_SUSTAINABILITY

Specifications

CAS 312692-97-0
Density 1.015
Molecular Formula C8H9IZn
MDL Number MFCD01311451
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive; May form a precipitate
Synonym zinc 2+ ion 3,4-dimethylbenzen-1-ide iodide
Solubility Information Reacts with water.
InChI Key VNMVACXRZXUZIP-UHFFFAOYSA-M
SMILES CC1=CC=[C-]C=C1C.[Zn+2].[I-]
IUPAC Name zinc;1,2-dimethylbenzene-5-ide;iodide
Molecular Weight (g/mol) 297.444
PubChem CID 57377101
Formula Weight 297.44
Chemical Name or Material 3,4-Dimethylphenylzinc iodide, Packaged under Argon in resealable ChemSeal™ bottles
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.