missing translation for 'onlineSavingsMsg'
Learn More

3,4-Epoxytetrahydrofuran, 96%

Product Code. 11906181 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass Bottle
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11906181 50 g Glass Bottle 50g
10429242 1 g Glass Bottle 1g
10041731 10 g Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11906181 Supplier Thermo Scientific Acros Supplier No. 425530500
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 285-69-8 | C4H6O2 | 86.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 285-69-8
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD00800639
InChI Key AIUTZIYTEUMXGG-UHFFFAOYNA-N
Synonym 3,4-epoxytetrahydrofuran, 3,6-dioxabicyclo 3.1.0 hexane, 3,6-dioxa-bicyclo 3.1.0 hexane, acmc-1ccip, 3,4-epoxy-tetrahydrofuran, 3,4-diepoxytetrahydrofuran, 3,4-epoxy tetrahydrofuran
PubChem CID 67511
IUPAC Name 3,6-dioxabicyclo[3.1.0]hexane
SMILES C1OCC2OC12

Specifications

Color Yellow
Density 1.2370g/mL
Quantity 50 g
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4450 to 1.4490
Assay Percent Range 95% min. (GC)
Specific Gravity 1.237
Solubility Information Solubility in water: moderately soluble in water.
Formula Weight 86.09
Percent Purity 96%
Chemical Name or Material 3, 4-Epoxytetrahydrofuran
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3,4-Epoxytetrahydrofuran
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.