missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Formylphenyl)propionic acid, 96%

Product Code. 15406767
Change view
Click to view available options
Quantity:
250 mg
5 g
Unit Size:
250mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15406767 250 mg 250mg
15496757 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15406767 Supplier Thermo Scientific Alfa Aesar Supplier No. H33873.MD
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 34961-64-3 | C10H10O3 | 178.19 g/mol

3-(4-Formylphenyl)propionic acid is used as an intermediate in the manufacture of pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(4-Formylphenyl)propionic acid is used as an intermediate in the manufacture of pharmaceuticals.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34961-64-3
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.19
MDL Number MFCD08234824
InChI Key NRCCSDVEUWXOMG-UHFFFAOYSA-N
Synonym 3-4-formylphenyl propanoic acid, 3-4-formylphenyl propionic acid, 3-4-formyl-phenyl-propionic acid, 3-4-formylphenyl-propionic acid, 4-formyldihydrocinnamic acid, 4-formylbenzenepropanoic acid, benzenepropanoic acid,4-formyl, 3-4-formylphenyl propanoicacid
PubChem CID 19825241
IUPAC Name 3-(4-formylphenyl)propanoic acid
SMILES OC(=O)CCC1=CC=C(C=O)C=C1

Specifications

Melting Point ∼136°C (decomposition)
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 178.19
Percent Purity 96%
Chemical Name or Material 3-(4-Formylphenyl)propionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.