missing translation for 'onlineSavingsMsg'
Learn More

3,4-Hexanedione, 96%

Product Code. 10687621 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
250 mL
Packaging:
Glass bottle
Unit Size:
250mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10687621 25 mL Glass bottle 25mL
10471291 250 mL Glass bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10687621 Supplier Thermo Scientific Acros Supplier No. 293270250
Spotlight Offers
Use promo code 28038
View all promotions
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4437-51-8 | C6H10O2 | 114.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4437-51-8
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00010237
InChI Key KVFQMAZOBTXCAZ-UHFFFAOYSA-N
Synonym 3,4-hexanedione, bipropionyl, dipropionyl, diethyl diketone, 3,4-hexane dione, unii-4m4npr5vgd, 3,4-hexandione, fema no. 3168, ccris 6295, 4m4npr5vgd
PubChem CID 62539
IUPAC Name hexane-3,4-dione
SMILES CCC(=O)C(=O)CC

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point -10°C
Color Yellow
Density 0.9410g/mL
Boiling Point 130°C
Flash Point 29°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula C2H5C(O)C(O)C2H5
Refractive Index 1.4090 to 1.415
Quantity 25 mL
Beilstein 01, 790
Specific Gravity 0.941
Solubility Information Solubility in water: 127g/L in water (20°C). Other solubilities: soluble in 1,2-propylene glycol,ethanol,oils,and fats
Formula Weight 114.14
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 3, 4-Hexanedione, 96%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3,4-Hexanedione
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.