missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Isopropylphenyl)acrylic Acid, TRC

Product Code. 30333773
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30333773 50 mg 50mg
30344512 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30333773 Supplier Toronto Research Chemicals Supplier No. TRCB45183050MG
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(4-Isopropylphenyl)acrylic Acid
CAS 3368-21-6
Molecular Formula C12H14O2
Quantity 50 mg
Synonym Cinnamic acid, p-isopropyl- (7CI,8CI), 3-[4-(1-Methylethyl)phenyl]-2-propenoic acid, 3-(4-Isopropylphenyl)-2-propenoic acid, 4-Isopropylcinnamic acid, NSC 216, p-Isopropylcinnamic acid
InChI Formula InChI=1S/C12H14O2/c1-9(2)11-6-3-10(4-7-11)5-8-12(13)14/h3-9H,1-2H3,(H,13,14)/b8-5+
SMILES CC(C)c1ccc(\C=C\C(=O)O)cc1
IUPAC Name (E)-3-(4-propan-2-ylphenyl)prop-2-enoic acid
Molecular Weight (g/mol) 190.24
Formula Weight 190.0994
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.