missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Methoxyphenyl)-1-propylboronic acid pinacol ester, 97%

Product Code. 11442080
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. Quantity unitSize
11442080 1 g 1g
15457745 250 mg 250mg
11452080 5 g 5g
3 options
This item is not returnable. View return policy

Product Code. 11442080

Brand: Thermo Scientific Alfa Aesar H28557.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1073371-72-8 | C16H25BO3 | 276.183 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1073371-72-8
Molecular Formula C16H25BO3
Molecular Weight (g/mol) 276.183
MDL Number MFCD09953503
InChI Key ZJNJYHQBUIHZLN-UHFFFAOYSA-N
Synonym 3-4-methoxyphenyl-1-propylboronic acid pinacol ester, 2-3-4-methoxyphenyl propyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-4-methoxyphenyl propylboronic acid pinacol ester, 3-4-methoxyphenyl propylboronic acid,pinacol ester, 3-4-methoxyphenyl propylboronic acid, pinacol ester
PubChem CID 46739066
IUPAC Name 2-[3-(4-methoxyphenyl)propyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)CCCC2=CC=C(C=C2)OC

Specifications

Quantity 1 g
Formula Weight 276.19
Percent Purity 97%
Chemical Name or Material 3-(4-Methoxyphenyl)-1-propylboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.