missing translation for 'onlineSavingsMsg'
Learn More

3,4-(Methylenedioxy)aniline, 97%

Product Code. 10647342 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10647342 10 g Glass bottle 10g
10581811 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10647342 Supplier Thermo Scientific Acros Supplier No. 171470100
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14268-66-7 | C7H7NO2 | 137.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14268-66-7
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00005832
InChI Key XGNXYCFREOZBOL-UHFFFAOYSA-N
Synonym 3,4-methylenedioxy aniline, benzo d 1,3 dioxol-5-amine, 5-amino-1,3-benzodioxole, 3,4-methylenedioxyaniline, 4-amino-1,2-methylenedioxybenzene, 5-aminobenzodioxole, methylenedioxyaniline, 2h-1,3-benzodioxol-5-amine, 1-amino-3,4-methylenedioxybenzene, 1-amino-3,4-dimethylenedioxybenzene
PubChem CID 84310
IUPAC Name 1,3-benzodioxol-5-amine
SMILES C1OC2=C(O1)C=C(C=C2)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 38.0°C to 43.0°C
Color Black to Brown
Boiling Point 144.0°C (16.0 mmHg)
Flash Point 142°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 10 g
Solubility Information Solubility in water: slightly soluble
Formula Weight 137.14
Percent Purity 97%
Physical Form Low Melting Solid
Chemical Name or Material 3, 4-(Methylenedioxy)aniline, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 3,4-(Methylenedioxy)aniline
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.