missing translation for 'onlineSavingsMsg'
Learn More

3,4-(Methylenedioxy)benzeneboronic acid, 98%

Product Code. 11451257
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11451257 1 g 1g
11461257 5 g 5g
11471257 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11451257 Supplier Thermo Scientific Alfa Aesar Supplier No. B24217.03
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 94839-07-3 | C7H7BO4 | 165.939 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94839-07-3
Molecular Formula C7H7BO4
Molecular Weight (g/mol) 165.939
MDL Number MFCD01009695
InChI Key CMHPUBKZZPSUIQ-UHFFFAOYSA-N
Synonym 3,4-methylenedioxyphenylboronic acid, benzo d 1,3 dioxol-5-ylboronic acid, 3,4-methylenedioxy phenylboronic acid, 3,4-methylenedioxyphenyl boronic acid, 3,4-methylenedioxy benzeneboronic acid, 1,3-benzodioxole-5-boronic acid, 3,4-methylenedioxybenzeneboronic acid, 2h-1,3-benzodioxol-5-yl boronic acid, 2h-1,3-benzodioxol-5-yl-boranediol, 2h-1,3-benzodioxol-5-ylboronic acid
PubChem CID 2734371
IUPAC Name 1,3-benzodioxol-5-ylboronic acid
SMILES B(C1=CC2=C(C=C1)OCO2)(O)O

Specifications

Melting Point 239°C to 243°C
Quantity 1 g
Beilstein 5523347
Formula Weight 165.94
Percent Purity 98%
Chemical Name or Material 3,4-(Methylenedioxy)benzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.