missing translation for 'onlineSavingsMsg'
Learn More

3,4-(Methylenedioxy)benzyl alcohol, 98%

Product Code. 11410533
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410533 25 g 25g
11420533 100 g 100g
11430533 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11410533 Supplier Thermo Scientific Alfa Aesar Supplier No. A12661.14
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 495-76-1 | C8H8O3 | 152.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 495-76-1
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00005836
InChI Key BHUIUXNAPJIDOG-UHFFFAOYSA-N
Synonym piperonyl alcohol, piperonol, 1,3-benzodioxole-5-methanol, 3,4-methylenedioxybenzyl alcohol, benzo d 1,3 dioxol-5-ylmethanol, piperonylalcohol, 5-hydroxymethyl-1,3-benzodioxole, 1-hydroxymethyl-3,4-methylenedioxybenzene, benzo 1,3 dioxol-5-ylmethanol, 3,4-methylenedioxy phenylmethanol
PubChem CID 10322
IUPAC Name 1,3-benzodioxol-5-ylmethanol
SMILES C1OC2=C(O1)C=C(C=C2)CO

Specifications

Melting Point 50°C to 55°C
Boiling Point 156°C to 158°C (16 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 136113
Formula Weight 152.15
Percent Purity 98%
Chemical Name or Material 3,4-(Methylenedioxy)benzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.