missing translation for 'onlineSavingsMsg'
Learn More

3,4-(Methylenedioxy)benzyl isothiocyanate, 94%, Thermo Scientific™

Product Code. 11398465
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11398465 5 g 5g
11388465 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398465 Supplier Thermo Scientific Alfa Aesar Supplier No. L10501.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3,4-(Methylenedioxy)benzyl isothiocyanate is used as intermediates. Used in nucleophilic addition reactions.

Solubility
Hydrolyses in water.

Notes
Moisture Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from moisture, alocohol, amines, water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4430-47-1
Molecular Formula C9H7NO2S
Molecular Weight (g/mol) 193.22
MDL Number MFCD00041217
InChI Key PUJWRDBPAFJUJW-UHFFFAOYSA-N
Synonym 3,4-methylenedioxy benzyl isothiocyanate, 1,3-benzodioxol-5-ylmethyl isothiocyanate, 5-isothiocyanatomethyl-1,3-benzodioxole, 3,4-methylenedioxybenzyl isothiocyanate, 5-isothiocyanatomethyl-2h-1,3-benzodioxole, 5-isothiocyanatomethyl-benzo 1,3 dioxole, 5-isothiocyanatomethyl benzo d 1,3 dioxole, acmc-20angn, pubchem9883, akos bbr-002798
PubChem CID 2795371
IUPAC Name 5-(isothiocyanatomethyl)-1,3-benzodioxole
SMILES S=C=NCC1=CC=C2OCOC2=C1

Specifications

Density 1.32
Boiling Point 139°C to 140°C (1 mmHg)
Odor Pungent
Refractive Index 1.615
Quantity 5 g
UN Number UN2922
Beilstein 1428941
Sensitivity Moisture sensitive
Solubility Information Hydrolyses in water.
Formula Weight 193.23
Percent Purity 94%
Chemical Name or Material 3,4-(Methylenedioxy)benzyl isothiocyanate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.