missing translation for 'onlineSavingsMsg'
Learn More

3,4-(Methylenedioxy)benzylamine, 97%, Thermo Scientific™

Product Code. p-7026055
Click to view available options
Quantity:
25 g
Unit Size:
25g
This item is not returnable. View return policy

Product Code. 11484877

Brand: Thermo Scientific Alfa Aesar B21837.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Piperonylamine is a metabolite of dopamine analogs. It is a pharmaceutical intermediate.

Solubility
Sparingly soluble in water. (0.9 g/100 mL).

Notes
Air sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store at 2-8°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2620-50-0
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00005840
InChI Key ZILSBZLQGRBMOR-UHFFFAOYSA-N
Synonym piperonylamine, benzo d 1,3 dioxol-5-ylmethanamine, 3,4-methylenedioxybenzylamine, 1,3-benzodioxol-5-ylmethylamine, 1,3-benzodioxole-5-methanamine, 2h-1,3-benzodioxol-5-ylmethanamine, 3,4-methylenedioxy benzylamine, benzo-1,3-dioxole-5-methylamine, 1-1,3-benzodioxol-5-yl methanamine, 5-aminomethyl-1,3-benzodioxole
PubChem CID 75799
SMILES NCC1=CC=C2OCOC2=C1

Specifications

Density 1.214
Boiling Point 136°C to 137°C (11 mmHg)
Flash Point >110°C (230°F)
Odor Amine-like
Refractive Index 1.564
Quantity 25 g
UN Number UN2735
Sensitivity Air Sensitive
Solubility Information Sparingly soluble in water. (0.9g/100 mL).
IUPAC Name 1,3-benzodioxol-5-ylmethanamine
Formula Weight 151.17
Percent Purity 97%
Chemical Name or Material 3,4-(Methylenedioxy)benzylamine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.