missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Methylphenyl)-3-(trifluoromethyl)diaziridine, TRC

Product Code. 30478075
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478075 25 mg 25mg
30480949 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30478075 Supplier Toronto Research Chemicals Supplier No. TRCM32601025MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87736-82-1
Molecular Formula C9 H9 F3 N2
Molecular Weight (g/mol) 202.18
IUPAC Name 3-(4-methylphenyl)-3-(trifluoromethyl)diaziridine
SMILES Cc1ccc(cc1)C2(NN2)C(F)(F)F

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C9H9F3N2/c1-6-2-4-7(5-3-6)8(13-14-8)9(10,11)12/h2-5,13-14H,1H3
Formula Weight 202.0718
Chemical Name or Material 3-(4-Methylphenyl)-3-(trifluoromethyl)diaziridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.