missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Methylpiperazin-1-yl)aniline, 97%, Thermo Scientific™

Product Code. 10797875
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10797875 5 g Amber Glass Bottle 5g
10012364 250 mg Amber Glass Bottle 250mg
10108912 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10797875 Supplier Thermo Scientific Maybridge Supplier No. CC35914DE
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 148546-99-0
Molecular Formula C11H17N3
Molecular Weight (g/mol) 191.278
InChI Key RJGHJWKQCJAJEP-UHFFFAOYSA-N
Synonym 3-4-methylpiperazin-1-yl aniline, 3-4-methyl-1-piperazinyl aniline, 3-4-methylpiperazin-1-yl benzenamine, benzenamine, 3-4-methyl-1-piperazinyl, zlchem 128, acmc-20a0ig, ksc496g5r, 3-4-methyl-piperazin-1-ylaniline, 3-4-methylpiperazinyl phenylamine, 3-4-methylpiperazine-1-yl aniline
PubChem CID 11564613
IUPAC Name 3-(4-methylpiperazin-1-yl)aniline
SMILES CN1CCN(CC1)C2=CC(=CC=C2)N

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 191.28
Percent Purity 97%
Chemical Name or Material 3-(4-Methylpiperazin-1-yl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.