missing translation for 'onlineSavingsMsg'
Learn More

3-(4-methylpiperazine-1-carbonyl)phenylboronic acid, 97%, Thermo Scientific™

Product Code. 10347154
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10347154 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10347154 Supplier Thermo Scientific Maybridge Supplier No. MAY00009DA
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 883738-38-3
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C18H27BN2O3
MDL Number MFCD05864308
Quantity 1 g
Synonym 4-methylpiperazin-1-yl 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanone, 3-4-methyl-1-piperazinylcarbonyl benzeneboronic acid pinacol ester, 3-4-methylpiperazine-1-carbonyl phenylboronic acid, pinacol ester, n-4-methylpiperazine-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl benzamide, 3-4-methylpiperazine-1-carbonyl phenyl boronic acid pinacol ester, 1-methyl-4-3-tetramethyl-1,3,2-dioxaborolan-2-yl benzoyl piperazine, 1-methyl-4-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoyl piperazine, n-4-methylpiperazine-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, amtb046
InChI Key ZYDMNNQZEUSDGG-UHFFFAOYSA-N
SMILES CN1CCN(CC1)C(=O)C1=CC=CC(=C1)B1OC(C)(C)C(C)(C)O1
IUPAC Name (4-methylpiperazin-1-yl)-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanone
Molecular Weight (g/mol) 330.24
PubChem CID 2760029
Formula Weight 330.23
Percent Purity 97%
Chemical Name or Material 3-(4-Methylpiperazine-1-carbonyl)phenylboronic acid, pinacol ester
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.