missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Morpholinyl)propyl isothiocyanate, 96%

Product Code. 11313386
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11313386 5 g 5g
11303386 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11313386 Supplier Thermo Scientific Alfa Aesar Supplier No. L12819.06
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 32813-50-6 | C8H14N2OS | 186.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32813-50-6
Molecular Formula C8H14N2OS
Molecular Weight (g/mol) 186.27
MDL Number MFCD00041222
InChI Key BCEFDMYFAAAFPE-UHFFFAOYSA-N
Synonym 3-morpholinopropyl isothiocyanate, 4-3-isothiocyanatopropyl morpholine, 3-4-morpholino propyl isothiocyanate, 3-morpholinopropylisothiocyanate, 3-morpholin-4-ylpropanisothiocyanate, 3-4-morpholinyl propyl isothiocyanate, 3-morpholin-4-yl propylisothiocyanate, 4-3-isothiocyanato-propyl-morpholine, 4-3-isothiocyanatopropyl morpholine #, morpholine,4-3-isothiocyanatopropyl
PubChem CID 141745
IUPAC Name 4-(3-isothiocyanatopropyl)morpholine
SMILES S=C=NCCCN1CCOCC1

Specifications

Boiling Point 182°C to 183°C (13 mmHg)
Refractive Index 1.5305
Quantity 5 g
UN Number UN2922
Beilstein 879217
Sensitivity Moisture sensitive
Formula Weight 186.28
Percent Purity 96%
Chemical Name or Material 3-(4-Morpholinyl)propyl isothiocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.