missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Nitrophenyl)adamantane-1-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10384772
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10384772 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10384772 Supplier Thermo Scientific Maybridge Supplier No. RJC00957DA
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 7123-76-4
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C17H19NO4
MDL Number MFCD00188065
Quantity 1 g
Synonym 3-4-nitrophenyl adamantane-1-carboxylic acid, maybridge1_006678, 3-4-nitrophenyl adamantanecarboxylic acid, 3-4-nitrophenyl-1-adamantanecarboxylic acid, tricyclo 3.3.1.13,7 decane-1-carboxylicacid, 3-4-nitrophenyl, tricyclo 3.3.1.1∼3,7∼ decane-1-carboxylic acid, 3-4-nitrophenyl
InChI Key JGYFFTQRRDPOIR-UHFFFAOYNA-N
SMILES OC(=O)C12CC3CC(C1)CC(C3)(C2)C1=CC=C(C=C1)[N+]([O-])=O
IUPAC Name 3-(4-nitrophenyl)adamantane-1-carboxylic acid
Molecular Weight (g/mol) 301.34
PubChem CID 2737411
Formula Weight 301.34
Percent Purity 97%
Chemical Name or Material 3-(4-Nitrophenyl)adamantane-1-carboxylic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.