missing translation for 'onlineSavingsMsg'
Learn More

3,4-Pyridinedicarboximide, 97%, Thermo Scientific™

Product Code. 10576104 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10576104 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10576104 Supplier Thermo Scientific Acros Supplier No. 425630050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 4664-01-1
CAS Min % 96.0
CAS Max % 100.0
Melting Point 232°C to 235°C
Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C7H4N2O2
MDL Number MFCD00013439
Quantity 5 g
Synonym 3,4-pyridinedicarboximide, cinchomeronimide, 3,4-pyridinecarboximide, 1h-pyrrolo 3,4-c pyridine-1,3 2h-dione, 2h-pyrrolo 3,4-c pyridine-1,3-dione, pyrrolo 3,4-c pyridine-1,3-dione, cinchomeronic acid imide, 4-azaphthalimid, pyridine-3,4-dicarboximide, 1h,2h,3h-pyrrolo 3,4-c pyridine-1,3-dione
InChI Key SJSABZBUTDSWMJ-UHFFFAOYSA-N
SMILES O=C1NC(=O)C2=CN=CC=C12
IUPAC Name pyrrolo[3,4-c]pyridine-1,3-dione
Molecular Weight (g/mol) 148.12
PubChem CID 72926
Formula Weight 148.12
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3, 4-Pyridinedicarboximide
Visa mer Visa mindre
compliance-icons
Produktbeteckning
  • 3,4-Pyridinedicarboximide
Signalord
  • Warning
Riskkategorier
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Faroangivelser
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Skyddsangivelse
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.