missing translation for 'onlineSavingsMsg'
Learn More

3,4-Pyridinedicarboxylic acid, 97%, Thermo Scientific Chemicals

Product Code. 10193610 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10193610 5 g Glass bottle 5g
10685182 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10193610 Supplier Thermo Scientific Acros Supplier No. 131890050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 490-11-9
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00006392
InChI Key MUYSADWCWFFZKR-UHFFFAOYSA-N
Synonym 3,4-pyridinedicarboxylic acid, cinchomeronic acid, chinchomeronic acid, 3,4-pyridinedicarboxylicacid, pyridine-3,4-dicarboxylicacid, unii-7389uzw5dy, 3,4-pyridindicarbons, cinchomeronsaure, pubchem2419, 3,4-dicarboxypyridine
PubChem CID 10273
ChEBI CHEBI:46860
IUPAC Name pyridine-3,4-dicarboxylic acid
SMILES OC(=O)C1=CC=NC=C1C(O)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 262°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Beilstein 22, 155
Merck Index 15, 2286
Solubility Information Solubility in water: .. Other solubilities: sparingly soluble in hot alcohol,ether and benzen,insoluble in chloroform
Formula Weight 167.12
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material 3, 4-Pyridinedicarboxylic acid, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 3,4-Pyridinedicarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.