missing translation for 'onlineSavingsMsg'
Learn More

3-[(4-pyridylmethyl)amino]propanenitrile, Thermo Scientific™

Product Code. 10476794
Change view
Click to view available options
Quantity:
5 g
Packaging:
Amber Glass Bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10476794 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10476794 Supplier Thermo Scientific Maybridge Supplier No. DSHS00705DE
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 648409-30-7
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C9H11N3
Quantity 5 g
Synonym 3-4-pyridylmethyl amino propanenitrile, 3-pyridin-4-ylmethyl amino propanenitrile, 3-pyridin-4-yl methyl amino propanenitrile, 3-pyridin-4-ylmethylamino propanenitrile, 3-4-pyridyl-methyl amino propanenitrile
InChI Key YBWTXEJAEKEKRW-UHFFFAOYSA-N
SMILES C1=CN=CC=C1CNCCC#N
IUPAC Name 3-(pyridin-4-ylmethylamino)propanenitrile
Molecular Weight (g/mol) 161.208
PubChem CID 2063970
Formula Weight 161.21
Percent Purity 97%
Chemical Name or Material 3-[(4-Pyridylmethyl)amino]propanenitrile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.