missing translation for 'onlineSavingsMsg'
Learn More

3-{[4-(tert-Butoxycarbonyl)piperazin-1-yl]methyl}benzoic acid, ≥97%, Thermo Scientific™

Product Code. 10491924
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10491924

Brand: Thermo Scientific Maybridge CC30401DE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 500013-38-7
Molecular Formula C17H24N2O4
Molecular Weight (g/mol) 320.389
MDL Number MFCD04115219
InChI Key CXJBKOKDFABTQO-UHFFFAOYSA-N
Synonym 3-4-tert-butoxycarbonyl piperazin-1-yl methyl benzoic acid, 1-3-carboxyphenylmethyl-4-boc piperazine, 1-boc-4-3-carboxy-benzyl-piperazine, 3-4-n-boc-piperazin-1-yl methylbenzoic acid, 1-tert-butoxycarbonyl-4-3-carboxybenzyl piperazine, tert-butyl 4-3-carboxybenzyl piperazine-1-carboxylate, 3-4-tert-butoxy carbonyl piperazin-1-yl methyl benzoic acid, 3-4-tert-butoxycarbonyl piperazin-1-yl methyl benzoicacid, 1-piperazinecarboxylicacid, 4-3-carboxyphenyl methyl-, 1-1,1-dimethylethyl ester
PubChem CID 2795521
IUPAC Name 3-[[4-[(2-methylpropan-2-yl)oxycarbonyl]piperazin-1-yl]methyl]benzoic acid
SMILES CC(C)(C)OC(=O)N1CCN(CC1)CC2=CC=CC(=C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 320.39
Percent Purity 97%
Chemical Name or Material 3-{[4-(tert-Butoxycarbonyl)piperazin-1-yl]methyl}benzoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.