missing translation for 'onlineSavingsMsg'
Learn More

3-[4-(Trifluoromethyl)phenyl]-1H-pyrazole-4-carbaldehyde, Thermo Scientific™

Product Code. 10374962
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10374962 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10374962 Supplier Thermo Scientific Maybridge Supplier No. SP01494DA
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 306936-65-2
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C11H7F3N2O
MDL Number MFCD02677691
Quantity 1 g
Synonym 3-4-trifluoromethyl phenyl-1h-pyrazole-4-carbaldehyde, 3-4-trifluoromethyl phenyl-1h-pyrazole-4-carboxaldehyde, 5-4-trifluoromethyl phenyl-1h-pyrazole-4-carbaldehyde, 3-4-trifluoromethylphenyl-4-pyrazolecarboxaldehyde, 3-4-trifluoromethyl phenyl pyrazole-4-carbaldehyde, 5-4-trifluoromethyl-phenyl-1h-pyrazole-4-c arbaldehyde, 5-4-trifluoromethyl phenyl-1h-pyrazole-4-carboxaldehyde, 1h-pyrazole-4-carboxaldehyde,3-4-trifluoromethyl phenyl, 3-4-trifluoromethyl phenyl-2h-pyrazole-4-carbaldehyde
InChI Key PMKPJAJMBQBTKE-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1C2=C(C=NN2)C=O)C(F)(F)F
IUPAC Name 5-[4-(trifluoromethyl)phenyl]-1H-pyrazole-4-carbaldehyde
Molecular Weight (g/mol) 240.185
PubChem CID 2781846
Formula Weight 240.18
Percent Purity 97%
Chemical Name or Material 3-[4-(Trifluoromethyl)phenyl]-1H-pyrazole-4-carbaldehyde
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.