missing translation for 'onlineSavingsMsg'
Learn More

3-[5-(1,5-Dioxo-5-(p-fluophenylpentyl]-4(S)-phenyl-2-oxazolidinone, TRC

Product Code. 30380057
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30380057 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380057 Supplier Toronto Research Chemicals Supplier No. TRCD486425100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 189028-93-1
Molecular Formula C20 H18 F N O4
Quantity 100 mg
InChI Formula InChI=1S/C20H18FNO4/c21-16-11-9-15(10-12-16)18(23)7-4-8-19(24)22-17(13-26-20(22)25)14-5-2-1-3-6-14/h1-3,5-6,9-12,17H,4,7-8,13H2/t17-/m1/s1
SMILES Fc1ccc(cc1)C(=O)CCCC(=O)N2[C@H](COC2=O)c3ccccc3
IUPAC Name 1-(4-fluorophenyl)-5-[(4S)-2-oxo-4-phenyl-1,3-oxazolidin-3-yl]pentane-1,5-dione
Molecular Weight (g/mol) 355.3596
Formula Weight 355.122
Percent Purity >95%
Chemical Name or Material 3-[5-(1,5-Dioxo-5-(p-fluophenylpentyl]-4(S)-phenyl-2-oxazolidinone
Grade HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.