missing translation for 'onlineSavingsMsg'
Learn More

3,5,7-Trimethyladamantane-1-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10160912
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10160912 250 mg Amber Glass Bottle 250mg
10384702 1 g Amber Glass Bottle 1g
10386494 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10160912 Supplier Thermo Scientific Maybridge Supplier No. BTB12973CB
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 15291-66-4
Molecular Formula C14H22O2
Molecular Weight (g/mol) 222.33
MDL Number MFCD00187581
InChI Key DHIIUPVOLLOORM-UHFFFAOYSA-N
Synonym 3,5,8-trimethyladamantanecarboxylic acid, maybridge4_000576, 1,3,5-trimethyladamantane-7-carboxylic acid, 3,5,7-trimethyl adamantane-1-carboxylic acid, 1s,3r,5s,7s-3,5,7-trimethyladamantane-1-carboxylic acid, 3r,5s,7s-3,5,7-trimethyladamantane-1-carboxylic acid, tricyclo 3.3.1.13,7 decane-1-carboxylicacid, 3,5,7-trimethyl, tricyclo 3.3.1.1∼3,7∼ decane-1-carboxylic acid, 3,5,7-trimethyl
PubChem CID 2802268
IUPAC Name 3,5,7-trimethyladamantane-1-carboxylic acid
SMILES CC12CC3(C)CC(C)(C1)CC(C2)(C3)C(O)=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 222.33
Percent Purity 97%
Chemical Name or Material 3, 5, 7-Trimethyladamantane-1-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.