missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxybenzhydrazide, 98+%

Product Code. 11470184
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11470184

Brand: Thermo Scientific Alfa Aesar A13619.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7466-54-8 | C8H10N2O2 | 166.18 g/mol

2-Methoxybenzhydrazide is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methoxybenzhydrazide is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7466-54-8
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00014755
InChI Key QMGXWNSSMGAHCA-UHFFFAOYSA-N
Synonym 2-methoxybenzhydrazide, 2-methoxybenzoic acid hydrazide, o-methoxybenzohydrazide, o-methoxybenzoylhydrazine, 2-methoxybenzoylhydrazide, 2-methoxybenzoylhydrazine, o-anisohydrazide, hydrazine, 2-methoxybenzoyl, o-methoxybenzoylhydrazide, o-methoxybenzoic acid hydrazide
PubChem CID 24051
IUPAC Name 2-methoxybenzohydrazide
SMILES COC1=CC=CC=C1C(=O)NN

Specifications

Melting Point 78°C to 82°C
Quantity 250 g
Beilstein 2208367
Solubility Information Slightly soluble in water.
Formula Weight 166.18
Percent Purity ≥98%
Chemical Name or Material 2-Methoxybenzhydrazide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.