missing translation for 'onlineSavingsMsg'
Learn More

3-(5-Bromo-2-thienyl)-5-methyl-1,2,4-oxadiazole, 97%, Thermo Scientific™

Product Code. 10435502
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10435502 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10435502 Supplier Thermo Scientific Maybridge Supplier No. CC19710DA
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 180530-13-6
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C7H5BrN2OS
MDL Number MFCD03407331
Quantity 1 g
Synonym 3-5-bromo-2-thienyl-5-methyl-1,2,4-oxadiazole, 3-5-bromothiophen-2-yl-5-methyl-1,2,4-oxadiazole, 3-2-bromothien-5-yl-5-methyl-1,2,4-oxadiazole, 1,2,4-oxadiazole,3-5-bromo-2-thienyl-5-methyl, 5-bromo-2-5-methyl 1,2,4-oxadiazol-3-yl thiophene
InChI Key ZNWQLJYQXULFQB-UHFFFAOYSA-N
SMILES CC1=NC(=NO1)C2=CC=C(S2)Br
IUPAC Name 3-(5-bromothiophen-2-yl)-5-methyl-1,2,4-oxadiazole
Molecular Weight (g/mol) 245.094
PubChem CID 2776504
Formula Weight 245.1
Percent Purity 97%
Chemical Name or Material 3-(5-Bromo-2-thienyl)-5-methyl-1, 2, 4-oxadiazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.