missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ 3,5-DHBA

Product Code. 18742524
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18742524 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18742524 Supplier R&D Systems™ Supplier No. 4599/50
  • ONLINE EXCLUSIVE PRICE
    105.38 €
    94.50 € / 50mg
    Save 10.88 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective HCA1 agonist

Selective agonist of hydroxycarboxylic acid receptor 1 (HCA1, also known as GPR81) (EC50 ∽150 μM). Inhibits lipolysis in wild-type mouse adipocytes. Displays minimal activity at HCA2 (at concentrations >10 mM) and no activity at HCA3, free fatty acid receptor (FFAR) 2 or FFAR3.
TRUSTED_SUSTAINABILITY

Specifications

CAS 99-10-5
Quantity 50 mg
Product Line Tocris
Molecular Formula C7H6O4
Formula Weight 154.12
Purity >99%
Molecular Weight (g/mol) 154.12
Chemical Name or Material 3, 5-DHBA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.