missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-t-butyl-2-hydroxybenzaldehyde, TRC

Product Code. 30370129
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30370129 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30370129 Supplier Toronto Research Chemicals Supplier No. TRCD4281505G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 37942-07-7
Molecular Formula C15H22O2
Quantity 5 g
Synonym 2-Hydroxy-3,5-di-tert-butylbenzaldehyde, 3,5-tert-Butyl-2-hydroxybenzaldehyde, 3,5-Bis(1,1-dimethylethyl)-2-hydroxybenzaldehyde, 3,5-Bis(tert-butyl)salicylaldehyde, 3,5-Di-t-butyl-2-hydroxybenzaldehyde, 3,5-Di-t-butylsalicylaldehyde, 3,5-Di-tert-butylsalicylaldehyde, 3,5-Di-tert-butylsalicylic Aldehyde
InChI Formula InChI=1S/C15H22O2/c1-14(2,3)11-7-10(9-16)13(17)12(8-11)15(4,5)6/h7-9,17H,1-6H3
SMILES CC(C)(C)c1cc(C=O)c(O)c(c1)C(C)(C)C
Molecular Weight (g/mol) 234.33
Formula Weight 234.162
Chemical Name or Material 3,5-Di-t-butyl-2-hydroxybenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.