missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butyl-o-benzoquinone, 97%, Maybridge

Product Code. 10421854
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10421854 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10421854 Supplier Thermo Scientific Maybridge Supplier No. BTB10297EB
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 3383-21-9
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Molecular Formula C14H20O2
Linear Formula [(CH3)3C]2C6H2(=O)2
MDL Number MFCD00001647
Quantity 25 g
Synonym 3,5-di-tert-butyl-o-benzoquinone, 3,5-di-tert-butyl-1,2-benzoquinone, 3,5-di tert-butyl benzo-1,2-quinone, 3,5-cyclohexadiene-1,2-dione, 3,5-bis 1,1-dimethylethyl, 3,5-di-tert-butylcyclohexa-3,5-diene-1,2-dione, o-benzoquinone, 3,5-di-tert-butyl, 3,5-di-tert-butylbenzo-1,2-quinone, 3,5-di-t-butyl-1,2-benzoquinone, 3,5-ditert-butylbenzo-1,2-quinone, 3,5-bis tert-butyl cyclohexa-3,5-diene-1,2-dione
InChI Key NOUZOVBGCDDMSX-UHFFFAOYSA-N
SMILES CC(C)(C)C1=CC(=O)C(=O)C(=C1)C(C)(C)C
IUPAC Name 3,5-ditert-butylcyclohexa-3,5-diene-1,2-dione
Molecular Weight (g/mol) 220.312
PubChem CID 76915
Formula Weight 220.31
Percent Purity 97%
Chemical Name or Material 3, 5-Di-tert-butyl-o-benzoquinone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.