missing translation for 'onlineSavingsMsg'
Learn More

3,5-Diamino-1,2,4-triazole, 98%

Product Code. 11499807
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499807 5 g 5g
11409817 25 g 25g
11419817 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499807 Supplier Thermo Scientific Alfa Aesar Supplier No. B22775.06
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1455-77-2 | C2H5N5 | 99.097 g/mol

3,5-Diamino-1,2,4-triazole is used as an inhibitor of DNA synthesis. It also serves as an antitumor agents in the treatment of epigenetically-based diseases. It acts as a corrosion inhibitor for copper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Air sensitive. Incompatible with acids and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1455-77-2
Molecular Formula C2H5N5
Molecular Weight (g/mol) 99.097
MDL Number MFCD00005233
InChI Key PKWIYNIDEDLDCJ-UHFFFAOYSA-N
Synonym guanazole, 3,5-diamino-1,2,4-triazole, 4h-1,2,4-triazole-3,5-diamine, 3,5-diamino-1h-1,2,4-triazole, 3,5-diamino-s-triazole, s-triazole, 3,5-diamino, 1,2,4-triazolidine, 3,5-diimino, unii-i01twm5267, ccris 3745, 3-amino-1h-1,2,4-triazol-5-ylamine
PubChem CID 15078
ChEBI CHEBI:75425
IUPAC Name 1H-1,2,4-triazole-3,5-diamine
SMILES C1(=NC(=NN1)N)N

Specifications

Melting Point ∼205°C (decomposition)
Quantity 5 g
Beilstein 112467
Solubility Information Soluble in water.
Formula Weight 99.1
Percent Purity ≥98%
Chemical Name or Material 3,5-Diamino-1,2,4-triazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.