missing translation for 'onlineSavingsMsg'
Learn More

3',5'-Dibromo-2'-hydroxyacetophenone, 99%

Product Code. 11450537
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11450537 25 g 25g
11440537 5 g 5g
11460537 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11450537 Supplier Thermo Scientific Alfa Aesar Supplier No. B22413.14
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 22362-66-9 | C8H6Br2O2 | 293.942 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22362-66-9
Molecular Formula C8H6Br2O2
Molecular Weight (g/mol) 293.942
MDL Number MFCD00191992
InChI Key RCKHUOIKVIOMGZ-UHFFFAOYSA-N
Synonym 3',5'-dibromo-2'-hydroxyacetophenone, 1-3,5-dibromo-2-hydroxyphenyl ethanone, 1-3,5-dibromo-2-hydroxyphenyl ethan-1-one, 3´,5´-dibromo-2´-hydroxyacetophenone, 1-acetyl-3,5-dibromo-2-hydroxybenzene, acmc-1cp7q, 2-acetyl-4,6-dibromophenol, dibromooxochromenecarboxylicacid, 3/',5/'-dibromo-2/'-hydroxyacetophenone, ethanone,1-3,5-dibromo-2-hydroxyphenyl
PubChem CID 271762
IUPAC Name 1-(3,5-dibromo-2-hydroxyphenyl)ethanone
SMILES CC(=O)C1=CC(=CC(=C1O)Br)Br

Specifications

Melting Point 108°C to 111°C
Quantity 25 g
Formula Weight 293.95
Percent Purity 99%
Chemical Name or Material 3',5'-Dibromo-2'-hydroxyacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.