missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dicarboethoxy-2,6-dimethyl-1,4-dihydropyridine, TRC

Product Code. 17934285
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17934285 1 g 1g
17994835 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17934285 Supplier Toronto Research Chemicals Supplier No. TRCD4310301G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1149-23-1
Molecular Formula C13 H19 N O4
Molecular Weight (g/mol) 253.29
Synonym 1,4-Dihydro-2,6-dimethyl-3,5-pyridinedicarboxylic Acid Diethyl Ester, 1,4-Dihydro-3,5-dicarboxylic Acid Diethyl Ester-2,6-dimethylpyridine, 2,6-Dimethyl-1,4-dihydropyridine-3,5-dicarboxylic Acid Diethyl Ester, 2,6-Dimethyl-3,5-di(carboethoxy)-1,4-dihydropyridine, 2,6-Dimethyl-3,5-dicarbethoxy-1,4-dihydropyridine, 2,6-Dimethyl-3,5-diethoxycarbonyl-1,4-dihydropyridine, 3,5-Bis(ethoxycarbonyl)-1,4-dihydro-2,6-dimethylpyridine, 3,5-Bis(ethoxycarbonyl)-2,6-dimethyl-1,4-dihydropyridine, 3,5-Dicarbethoxy-2,6-dimethyl-1,4-dihydropyridine, 3,5-Dicarboethoxy-2,6-dimethyl-1,4-dihydropyridine, 3,5-Diethoxycarbonyl-1,4-dihydro-2,6-dimethylpyridine, 3,5-Diethoxycarbonyl-2,6-dimethyl-1,4-dihydropyridine, Diethone, Diethyl 1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate, Diethyl 2,6-dimethyl-1,4-dihydr
IUPAC Name diethyl 2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
SMILES CCOC(=O)C1=C(C)NC(=C(C1)C(=O)OCC)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C13H19NO4/c1-5-17-12(15)10-7-11(13(16)18-6-2)9(4)14-8(10)3/h14H,5-7H2,1-4H3
Formula Weight 253.1314
Chemical Name or Material 3,5-Dicarboethoxy-2,6-dimethyl-1,4-dihydropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.