missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichloro-2-hydroxybenzaldehyde hydrazone, 98%, Thermo Scientific™

Product Code. 11975661 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11975661 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11975661 Supplier Thermo Scientific Acros Supplier No. 438320050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 43002-22-8
CAS Min % 97.5
CAS Max % 100.0
Melting Point 149°C to 151°C
Color Brown to White
Elemental Analysis 39.98 to 42.03% C, 13.32 to 14.00% N
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C7H6Cl2N2O
MDL Number MFCD00221441
Quantity 5 g
Synonym 2,4-dichloro-6-hydrazinylmethylidene cyclohexa-2,4-dien-1-one, 2,4-dichloro-6-methanehydrazonoylphenol, benzaldehyde,3,5-dichloro-2-hydroxy-, hydrazone, 2,4-bis chloranyl-6-diazanylmethylidene cyclohexa-2,4-dien-1-one, 2,4-dichloro-6-hydrazinylmethylidene-1-cyclohexa-2,4-dienone
InChI Key HJIQERSFUMXSNW-UHFFFAOYSA-N
SMILES NNC=C1C=C(Cl)C=C(Cl)C1=O
IUPAC Name 2,4-dichloro-6-(hydrazinylmethylidene)cyclohexa-2,4-dien-1-one
Molecular Weight (g/mol) 205.04
PubChem CID 6807126
Formula Weight 205.04
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3, 5-Dichloro-2-hydroxybenzaldehyde hydrazone
Show More Show Less
compliance-icons
Product Identifier
  • 3,5-Dichloro-2-hydroxybenzaldehyde hydrazone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.