missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichloro-alpha-toluenesulfonyl chloride, 96%

Product Code. 11490260
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490260 250 mg 250mg
11400270 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11490260 Supplier Thermo Scientific Alfa Aesar Supplier No. H50707.MD
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 163295-70-3 | C7H5Cl3O2S | 259.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 163295-70-3
Molecular Formula C7H5Cl3O2S
Molecular Weight (g/mol) 259.53
MDL Number MFCD01631928
InChI Key GZMJRPMBVICCFL-UHFFFAOYSA-N
Synonym 3,5-dichlorophenyl methanesulfonyl chloride, 3,5-dichloro-phenyl-methanesulfonyl chloride, acmc-1c80o, 3,5-dichloro-, a-toluenesufonyl chloride, 3,5-dichlorophenyl methyl chlorosulfone, 3,5-dichlorophenyl methanesulfonylchloride, 3,5-dichloro-alpha-toluenesulfonyl chloride, 3,5-dichlorophenyl methane-sulfonyl chloride, benzenemethanesulfonylchloride, 3,5-dichloro, 3,5-dichlorophenyl methyl sulfonyl chloride
PubChem CID 4993270
IUPAC Name (3,5-dichlorophenyl)methanesulfonyl chloride
SMILES ClC1=CC(CS(Cl)(=O)=O)=CC(Cl)=C1

Specifications

Quantity 250 mg
UN Number UN3096
Formula Weight 259.54
Percent Purity 96%
Chemical Name or Material 3,5-Dichloro-alpha-toluenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.