missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorobenzenesulfonyl chloride, 97%

Product Code. 11494453
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11494453 1 g 1g
11404463 5 g 5g
11414463 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494453 Supplier Thermo Scientific Alfa Aesar Supplier No. A11003.03
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 705-21-5 | C6H3Cl3O2S | 245.50 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 705-21-5
Molecular Formula C6H3Cl3O2S
Molecular Weight (g/mol) 245.50
MDL Number MFCD00051698
InChI Key RJSQINMKOSOUGT-UHFFFAOYSA-N
Synonym 3,5-dichlorobenzene-1-sulfonyl chloride, 3,5-dichlorobenzenesulphonyl chloride, 3,5-dichlorobenzenesulfonylchloride, 3,5-dichloro-benzenesulfonyl chloride, 3,5-dichlorophenyl chlorosulfone, 3,5-dichlorophenylsulfonyl chloride, benzenesulfonyl chloride, 3,5-dichloro, pubchem5120, acmc-209oey, ksc377a3t
PubChem CID 522388
IUPAC Name 3,5-dichlorobenzenesulfonyl chloride
SMILES ClC1=CC(=CC(Cl)=C1)S(Cl)(=O)=O

Specifications

Melting Point 30°C to 33°C
Boiling Point 158°C to 160°C (15 mmHg)
Flash Point >110°C (230°F)
Quantity 1 g
UN Number UN3261
Beilstein 1528657
Sensitivity Moisture sensitive
Formula Weight 245.51
Percent Purity 97%
Chemical Name or Material 3,5-Dichlorobenzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.