missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dichlorobenzoic-d3 Acid, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 3,5-Dichlorobenzoic-d3 Acid |
| CAS | 1219803-99-2 |
| Molecular Formula | C7 D3 H Cl2 O2 |
| Quantity | 0.1 g |
| Synonym | 3,5-Dichlorobenzoic-d3 Acid, 3,5-Dichlorobenzoic Acid-d3, 3,5-Dichlorobenzoic-2,4,6-d3 Acid, 3,5-Dichlorobenzoic Acid-D3 |
| InChI Formula | InChI=1 S/C7H4Cl2O2/c8-5-1-4(7(10)11)2-6(9)3-5/h1-3 H,(H,10,11)/i1D,2 D,3 D |
| SMILES | [2 H]c1c(Cl)c([2 H])c(C(=O)O)c([2 H])c1Cl |
| IUPAC Name | 3,5-dichloro-2,4,6-trideuteriobenzoic acid |
| Molecular Weight (g/mol) | 194.03 |
| Formula Weight | 192.978 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction