missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorophenyl isocyanate, 97%

Product Code. 11430431
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11430431 25 g 25g
11420431 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11430431 Supplier Thermo Scientific Alfa Aesar Supplier No. A19991.14
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 34893-92-0 | C7H3Cl2NO | 188.007 g/mol

3,5-Dichlorophenyl isocyanate is used as an intermediate for medicine and in iprodione. It is also useful for proteomic research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dichlorophenyl isocyanate is used as an intermediate for medicine and in iprodione. It is also useful for proteomic research.

Solubility
Hydrolysis with water.

Notes
Light and moisture sensitive. Incompatible with water, alcohols, strong bases, amines, acids and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34893-92-0
Molecular Formula C7H3Cl2NO
Molecular Weight (g/mol) 188.007
MDL Number MFCD00013859
InChI Key XEFUJGURFLOFAN-UHFFFAOYSA-N
Synonym 3,5-dichlorophenyl isocyanate, 3,5-dichlorophenylisocyanate, benzene, 1,3-dichloro-5-isocyanato, 3,5-dichlorophenyl isocyantae, 1,3-dichloro-5-isocyanato-benzene, 3,5-dichlorobenzenisocyanate, 3,5dichlorophenyl isocyanate, acmc-1ai47, ksc490c1l, 3,5-dichloro-phenylisocyanate
PubChem CID 94460
IUPAC Name 1,3-dichloro-5-isocyanatobenzene
SMILES C1=C(C=C(C=C1Cl)Cl)N=C=O

Specifications

Melting Point 28°C to 32°C
Density 1.38
Boiling Point 242°C to 244°C
Flash Point 134°C (273°F)
Quantity 25 g
UN Number UN2250
Beilstein 510282
Sensitivity Moisture sensitive
Solubility Information Hydrolysis with water.
Formula Weight 188.01
Percent Purity 97%
Chemical Name or Material 3,5-Dichlorophenyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.