missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorophenylacetic acid, 95%

Product Code. 15499918
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15499918 5 g 5g
15409928 250 mg 250mg
15489918 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15499918 Supplier Thermo Scientific Alfa Aesar Supplier No. H63293.06
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 51719-65-4 | C8H6Cl2O2 | 205.034 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51719-65-4
Molecular Formula C8H6Cl2O2
Molecular Weight (g/mol) 205.034
MDL Number MFCD01764671
InChI Key RERINLRFXYGZEE-UHFFFAOYSA-N
Synonym 2-3,5-dichlorophenyl acetic acid, 3,5-dichlorophenylacetic acid, benzeneacetic acid, 3,5-dichloro, 3,5-dichlorobenzeneacetic acid, 3,5-dichlorophenyl acetic acid, 3,5-dichloro-phenyl-acetic acid, 2-3,5-dichlorophenyl-acetic acid, 3,5-dichlorophenylaceticacid, pubchem24116, 3,5-dichlorphenylacetic acid
PubChem CID 6452487
IUPAC Name 2-(3,5-dichlorophenyl)acetic acid
SMILES C1=C(C=C(C=C1Cl)Cl)CC(=O)O

Specifications

Quantity 5 g
Formula Weight 205.03
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3,5-Dichlorophenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.