missing translation for 'onlineSavingsMsg'
Learn More

3,5-dichloropyridazine, Thermo Scientific™

Product Code. 10585174
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10585174 1 g Amber Glass Bottle 1g
10142894 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10585174 Supplier Thermo Scientific Maybridge Supplier No. MO08501DA
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1837-55-4
Molecular Formula C4H2Cl2N2
Molecular Weight (g/mol) 148.97
MDL Number MFCD10698048
InChI Key JZSAUQMXKHBZEO-UHFFFAOYSA-N
PubChem CID 19959687
IUPAC Name 3,5-dichloropyridazine
SMILES ClC1=CC(Cl)=NN=C1

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Synonym pyridazine, 3,5-dichloro, 3,5-dichloro-pyridazine, pyridazine,3,5-dichloro, 3,5-dichlor-pyridazin, acmc-209yx3, ksc538k8n
Formula Weight 148.98
Percent Purity 97%
Chemical Name or Material 3, 5-dichloropyridazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.