missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorosulfanilamide, 97%, Thermo Scientific™

Product Code. 11341447
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11341447 25 g 25g
11331447 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11341447 Supplier Thermo Scientific Alfa Aesar Supplier No. L01939.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22134-75-4
Molecular Formula C6H6Cl2N2O2S
Molecular Weight (g/mol) 241.086
MDL Number MFCD00014784
InChI Key DVZMRTJKNJKEGV-UHFFFAOYSA-N
Synonym 3,5-dichlorosulfanilamide, benzenesulfonamide, 4-amino-3,5-dichloro, 4-amino-3,5-dichlorobenzene-1-sulfonamide, sulfanilamide, 3,5-dichloro, 4-amino-3,5-dichloro-benzenesulfonamide, 3,5-dichlorosulphanilamide, 1zge, acmc-1clhm, sulfanilamide,5-dichloro, aronis24244
PubChem CID 89607
IUPAC Name 4-amino-3,5-dichlorobenzenesulfonamide
SMILES C1=C(C=C(C(=C1Cl)N)Cl)S(=O)(=O)N

Specifications

Melting Point 206°C to 208°C
Quantity 25 g
Beilstein 2727109
Formula Weight 241.1
Percent Purity 97%
Chemical Name or Material 3,5-Dichlorosulfanilamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.