missing translation for 'onlineSavingsMsg'
Learn More

3,5-Diethyl-1,2-dihydro-1-phenyl-2-n-propylpyridine, tech. 75%, Thermo Scientific™

Product Code. 15418125
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15418125 50 g 50g
15408125 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15418125 Supplier Thermo Scientific Alfa Aesar Supplier No. H32538.18
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34562-31-7
Molecular Formula C18H25N
Molecular Weight (g/mol) 255.41
MDL Number MFCD00071789
InChI Key IAAASXBHFUJLHW-UHFFFAOYNA-N
Synonym 3,5-diethyl-1-phenyl-2-propyl-1,2-dihydropyridine, 3,5-diethyl-1,2-dihydro-1-phenyl-2-propylpyridine, pdhp, pyridine, 3,5-diethyl-1,2-dihydro-1-phenyl-2-propyl, 1-phenyl-3,5-diethyl-2-propyl-1,2-dihydropyridine, dsstox_cid_24923, dsstox_rid_80589, dsstox_gsid_44923, 3,5-diethyl-1,2-dihydro-1-phenyl-2-n-propylpyridine, 3,5-diethyl-1,2-dihydro-1-phenyl-2-propyl pyridine
PubChem CID 118703
IUPAC Name 3,5-diethyl-1-phenyl-2-propyl-2H-pyridine
SMILES CCCC1N(C=C(CC)C=C1CC)C1=CC=CC=C1

Specifications

Density 0.97
Boiling Point 125°C (0.5 mmHg)
Flash Point >100°C (212°F)
Refractive Index 1.5675
Quantity 50 g
Formula Weight 255.4
Percent Purity 75%
Grade Technical
Chemical Name or Material 3,5-Diethyl-1,2-dihydro-1-phenyl-2-n-propylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.